Forceps with guide pin
Curved, with guide pin, 200 mm
目录编号 RSGA312.200
供应商: RSG
Jump to:
Product Details & Documents
Stainless steel, anti-magnetic. Particularly suitable for staining applications.
- With curved, extra fine tips
- Corrosion resistant
规格
- Type:Curved, with guide pin
- 长度:200 mm