Spoon spatulas, micro
210 mm, 7 mm
目录编号 RSGA627.210
供应商: RSG
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
- With a micro spoon at one end and a micro spatula at the other
- Excellent chemical resistance
- Excellent thermal resistance
- Quick and easy to clean
Documents
规格
- 长度:210 mm
- 刀片宽度:7 mm