Staining trough
Staining trough tray, for 20 slides, 76×26 mm
目录编号 BRND474305
供应商: BRAND
Jump to:
Product Details & Documents
PMP, transparent.
Trough comes with two lids: one to help protect against evaporation when not in use; one with a handle slot facilitating the staining process.
PP tray with fold down handle can accomodate 20 slides. The close fitting lid helps protect against evaporation.
Ordering information: Trough and tray (insert) have to be ordered separately.
规格
- 说明:Staining trough tray, for 20 slides, 76×26 mm