Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
200 mm, 40 mm, 5 mm, 9×5 mm
目录编号 BOCH3345
供应商:   BOCHEM
Double spatulas, micro
目录编号 BOCH3345
供应商:  BOCHEM
Double spatulas, micro
Double spatulas, micro
200 mm, 40 mm, 5 mm, 9×5 mm
Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Double spatulas for a wide range of uses.
  • One end with micro spoon or scoop and one end with micro spatula
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Documents
规格
  • 长度:
    200 mm
  • 刀片宽度:
    5 mm
  • 线圈尺寸:
    9×5 mm
  • 刀片长度:
    40 mm