Wire baskets, square and rectangular
150×200×150 mm
目录编号 BOCH10027
供应商: BOCHEM
Jump to:
Product Details & Documents
These stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
- Spot-welded grid 8×8×1 mm
- Reinforced edges
- Very good chemical and temperature resistance
规格
- 宽度×深度×高度:150×200×150 mm