Digital Solutions
Applications
Support
About
China
Orders Link
ContactUsLinkComponent
搅拌棒
Stand rods, with M10 thread
Stand rods, with M10 thread
12 mm, 1000 mm
BOCH5133
:
BOCHEM
Stand rods, with M10 thread
BOCH5133
:
BOCHEM
Stand rods, with M10 thread
12 mm, 1000 mm
1 * 1 件
-
+
Aluminium, bare.
Lightweight, tough
With M10 thread
直径:
12 mm
长度:
1000 mm