Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
订单输入
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
搅拌棒
Retort rods, without thread
Retort rods, without thread
Print
Retort rods, without thread
15, 13 mm, 1500 mm
Catalog # BOCH5185
Supplier:
BOCHEM
Retort rods, without thread
Catalog # BOCH5185
Supplier:
BOCHEM
Expand image
Return to Top
Retort rods, without thread
15, 13 mm, 1500 mm
Specifications
Add to cart
Jump to:
Specifications
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 件
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
Stainless steel 18/10.
Rods for stand bases
Without thread
Specifications
直径:
13 mm
长度:
1500 mm
顺序:
15