订单输入
China
Orders LinkContactUsLinkComponent
 

碳酸钠 ≥97%, TECHNICAL

4114, Plastic bucket, 5 kg
目录编号 VWRC27754.360
刺激性
供应商:   VWR Chemicals
碳酸钠 ≥97%, TECHNICAL
目录编号 VWRC27754.360
供应商:  VWR Chemicals
CAS号:  497-19-8
碳酸钠 ≥97%, TECHNICAL
碳酸钠 ≥97%, TECHNICAL

4114, Plastic bucket, 5 kg

Jump to:
CAS号:  497-19-8
Price
¥805.00
Log in 查看库存
1 * 5 千克
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
同义词:  纯碱
  • CAS号:497-19-8
  • 化学式:Na₂CO₃
  • MDL 编号:MFCD00003494
  • Molecular Weight:105.99 g/mol
  • 默克索引:14,08596
  • 沸点:1600 °C (1013 hPa)
  • 熔点:854 °C
  • 密度:2,53 g/cm³ (20 °C)
  • 储存温度:Ambient
  • EINECS:207-838-8
  • REACH:01-2119485498-19
规格

规格测试结果

  • Assay (on anhydrous substance):
    Min. 97 %

规格

  • 包装类型:
    Plastic bucket
  • 包装规格:
    5 kg
  • 顺序:
    4114

规格

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.