Orders LinkContactUsLinkComponent
Test kits, digital titration method
Test kits, digital titration method
17, Total hardness HR: EDTA method, 100 - 4000 mg/l, 100 Tests
Catalog # HACH2448100
Supplier:   HACH
Test kits, digital titration method
Catalog # HACH2448100
Supplier:  HACH
CAS Number:  
Test kits, digital titration method
Test kits, digital titration method
17, Total hardness HR: EDTA method, 100 - 4000 mg/l, 100 Tests
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 Tests
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • 包装规格:
    100 Tests
  • 说明:
    Total hardness HR: EDTA method
  • 顺序:
    17
  • 范围:
    100 - 4000 mg/l
Specifications