跳转到 页眉
跳转到 主要内容
跳转到 页脚
高级搜索
SDS
Certificates
制造商数据
0
¥0.00
订单输入
登录
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
您的账户
Cart
China
Order Entry
Order Status
您的账户
Cart
Orders Link
ContactUsLinkComponent
Home
搅拌棒
Retort rods, without thread
Retort rods, without thread
Print
Retort rods, without thread
14, 13 mm, 1250 mm
目录编号 BOCH5184
供应商:
BOCHEM
Retort rods, without thread
目录编号 BOCH5184
供应商:
BOCHEM
展开图片
Return to Top
Retort rods, without thread
14, 13 mm, 1250 mm
规格
Add to cart
Jump to:
规格
Price
询价
Log in
查看库存
规格
1 * 1 件
-
+
询价
Other Product Family Options Available
Return Policy
Product Details & Documents
Stainless steel 18/10.
Rods for stand bases
Without thread
规格
直径:
13 mm
长度:
1250 mm
顺序:
14