二氟氢化铵 ≥97%, TECHNICAL
4024, Plastic bottle for solids, 1 kg
供应商: VWR Chemicals
Jump to:
Product Details & Documents
同义词: 双氟氨, 蚀刻粉
- CAS号:1341-49-7
- 化学式:NH₄HF₂
- MDL 编号:MFCD00012018
- Molecular Weight:57.04 g/mol
- 默克索引:13,00498
- 熔点:127 °C
- 密度:0,8 g/cm³ (20 °C)
- 储存温度:Ambient
- EINECS:215-676-4
- UN:1727
- ADR:8,II
规格
规格测试结果
- Assay:Min. 97 %
规格
- 包装类型:Plastic bottle for solids
- 包装规格:1 kg
- 顺序:4024
规格
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



