Orders LinkContactUsLinkComponent
Test kits, digital titration method
Test kits, digital titration method
28, Silver nitrate digital titrator cartridge, 1,128 N, 1
目录编号 HACH1439701
供应商:   HACH
Test kits, digital titration method
目录编号 HACH1439701
供应商:  HACH
CAS号:  
Test kits, digital titration method
Test kits, digital titration method
28, Silver nitrate digital titrator cartridge, 1,128 N, 1
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
规格
规格
  • 包装规格:
    1
  • 说明:
    Silver nitrate digital titrator cartridge, 1,128 N
  • 顺序:
    28
规格