Orders LinkContactUsLinkComponent
Test kits, digital titration method
06, Total hardness HA-DT: EDTA method, 10 - 4000 mg/l, 1 KIT
目录编号 HACH2063600
供应商:   HACH
Test kits, digital titration method
目录编号 HACH2063600
供应商:  HACH
CAS号:  
Test kits, digital titration method
Test kits, digital titration method
06, Total hardness HA-DT: EDTA method, 10 - 4000 mg/l, 1 KIT
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 1 KIT
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
规格
规格
  • 包装规格:
    1 KIT
  • 说明:
    Total hardness HA-DT: EDTA method
  • 顺序:
    06
  • 范围:
    10 - 4000 mg/l
规格