China
Orders LinkContactUsLinkComponent
Manual microorganism identification reagents and medium, API®
Manual microorganism identification reagents and medium, API®
API® TDA reagents, 2×5 ml, 1 KIT
  BMER70402
Marketsource
 :   BIOMERIEUX
Manual microorganism identification reagents and medium, API®
  BMER70402
Marketsource
 :  
Manual microorganism identification reagents and medium, API®
API® TDA reagents, 2×5 ml, 1 KIT
 :  
1 * 1 KIT
API® Identification kits provide simple methodology for detection of various groups of organisms and also enzymes. It includes the evolution of international taxonomy, description of the new bacterial species and newly acquired bacteriology data.
  • Rapid and reliable method for bacterial identification
Documents
  • 包装规格:
    1 KIT
  • 说明:
    API® TDA reagents, 2×5 ml