Digital Solutions
Applications
Support
About
China
Orders Link
ContactUsLinkComponent
研磨机
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
07, Funnel, A 11.7, Use with liquid nitrogen, prevents splashing when pouring
IKAA3048700
:
IKA JANKE KUNKEL
Accessories for analytical Mill, A 11 basic
IKAA3048700
:
IKA JANKE KUNKEL
7
Accessories for analytical Mill, A 11 basic
07, Funnel, A 11.7, Use with liquid nitrogen, prevents splashing when pouring
1 * 1 件
-
+
说明:
Funnel, A 11.7
顺序:
07
用于:
Use with liquid nitrogen, prevents splashing when pouring