Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
01, 14/23 NS, 14/23 NS
目录编号 LENZ05000111
供应商:   Lenz Laborglas
Multiple adapters
目录编号 LENZ05000111
供应商:  Lenz Laborglas
Multiple adapters
Multiple adapters
01, 14/23 NS, 14/23 NS
Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two standard ground 29/32 sockets and one standard ground 29/32 cone. Side neck inclined upwards at an angle of 105°.
Documents
规格
  • 圆锥体大小:
    14/23 NS
  • 插座尺寸:
    14/23 NS
  • 顺序:
    01