Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
250 ml, 29/32 NS, -
目录编号 LENZ04004149
供应商:   Lenz Laborglas
Conical separating funnels
目录编号 LENZ04004149
供应商:  Lenz Laborglas
Conical separating funnels
Conical separating funnels
250 ml, 29/32 NS, -
Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.
Documents
规格
  • 插座尺寸:
    29/32 NS
  • 容量:
    250 ml
  • 分度:
    -