Buffer solutions in sachets
10,01, ±0,01, Sachet, 25×20 ml, 25
供应商: HANNA
Jump to:
Product Details & Documents
For calibration of pH meters.
- Colour coded label for easy pH buffer value identification
- Ideal for field use due to portability
- Light block packaging prevents oxidation from UV light
Traceable to NIST standard reference materials.
Documents
Certificates
按批号搜索规格
规格
- 包装类型:Sachet
- 包装规格:25
- pH精度:±0,01
- Pk Info:25×20 ml
- pH 值:10,01



