Digital Solutions
Applications
Support
About
China
Orders Link
ContactUsLinkComponent
Forceps
Forceps
Forceps
Forceps
Straight, pointed, 115 mm
RSGA613.115
:
RSG
Forceps
RSGA613.115
:
RSG
Expand image
Forceps
Straight, pointed, 115 mm
Add to cart
Unit of Measure
1 * 1 件
-
+
Stainless steel 18/10, anti-magnetic.
Documents
Datasheet
RSGA611.300_TDS_EN
Type:
Straight, pointed
长度:
115 mm