Forceps
Straight, pointed, 115 mm
目录编号 RSGA413.115
供应商: RSG
Jump to:
Product Details & Documents
Stainless steel, PTFE coated.
- Available in sharp or blunt design
- Very good chemical and thermal resistance
- Smooth surfaces prevent contamination and allow easy cleaning
规格
- Type:Straight, pointed
- 长度:115 mm