Accessories for analytical Mill, A 11 basic
04, Beater A11.3, Pulverising substances with a Mohs hardness of up to 9
目录编号 IKAA2983000
供应商: IKA JANKE KUNKEL
Jump to:
Product Details & Documents
规格
- 说明:Beater A11.3
- 顺序:04
- 用于:Pulverising substances with a Mohs hardness of up to 9