Forceps
Straight, pointed, 115 mm
目录编号 VITL67895
供应商: VITLAB
Jump to:
Product Details & Documents
PMP, white. Multipurpose forceps for a wide range of applications.
- Good chemical resistance
- Autoclavable
- Robust, pliable, self-sprung
规格
- Type:Straight, pointed
- 长度:115 mm