Orders LinkContactUsLinkComponent
Volumetric flasks, class B
07, 1000 ml, 24/29, 0,80 ± ml
  VITL676941
 :   VITLAB
Volumetric flasks, class B
  VITL676941
 :  VITLAB
Volumetric flasks, class B
Volumetric flasks, class B
07, 1000 ml, 24/29, 0,80 ± ml
1 * 1 件
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
  • 模拟器:
    24/29
  • 容量:
    1000 ml
  • 顺序:
    07
  • 误差:
    0,80 ± ml