Scoops
1500 ml, 350 mm
供应商: RSG
Jump to:
Product Details & Documents
Stainless steel. Manual scoops for collecting or measuring powders, pastes or granulates.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Robust design
Documents
Certificates
按批号搜索规格
- 容量:1500 ml
- 长度:350 mm