Scoops
450 ml, 250 mm
目录编号 RSGA758.075
供应商: RSG
Jump to:
Product Details & Documents
Stainless steel. Manual scoops for collecting or measuring powders, pastes or granulates.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Robust design
Documents
Certificates
按批号搜索规格
- 容量:450 ml
- 长度:250 mm