Containers with optional snap caps, Nunc™
for 12,5/20 and 30 ml containers, 自然色
目录编号 NUNC536617
Containers with optional snap caps, Nunc™
目录编号 NUNC536617
Jump to:
Product Details & Documents
PS, clear.
- Transparent polystyrene construction
- Versatile containers made of high-quality resins
- Snap caps available for all containers
- Cylindrical shape
- Non sterile
Ordering information: Caps must be ordered seperately.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
规格
- Type:for 12,5/20 and 30 ml containers
- 盖颜色:自然色
- Cat. No.:NUNC536617
- 环境认证:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact