Orders LinkContactUsLinkComponent
Modules, Nunc-Immuno™ LockWell™
27, MaxiSorp™ C8×12 modules, 深红色
目录编号 NUNC477089
Modules, Nunc-Immuno™ LockWell™
目录编号 NUNC477089
Modules, Nunc-Immuno™ LockWell™
Modules, Nunc-Immuno™ LockWell™
27, MaxiSorp™ C8×12 modules, 深红色
Price
询价
Log in 查看库存
1 * 60 件
Product Details & Documents
PS. Nunc-Immuno™ LockWell™ modules facilitate assays from large scale screening to single-well tests. Available with a choice of MaxiSorp™, PolySorp™, MediSorp™ and MultiSorp™ surfaces. All MaxiSorp™ modules are certified for binding homogeneity. Framed modules (external dimensions: 128×86 mm) fit all standard equipment.
  • Each well locks in the frame at the same height to prevent jamming in automated systems
  • Strips are easy to take out or put in the frame, without use of any special tools
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains; PolySorp™ surface has a high affinity for hydrophobic molecules; MediSorp™ has a surface chemistry intermediate between PolySorp™ and MaxiSorp™, providing low background signal with samples containing serum; MultiSorp™ surfaces have a high affinity to hydrophilic molecules.
Colour coded modules available on request, please visit website or contact your local Avantor sales office for further information.
规格
  • 颜色:
    深红色
  • 说明:
    MaxiSorp™ C8×12 modules
  • 顺序:
    27
Accessories
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • NUNC465404
    42
    LockWell™ frame
    Log in 查看库存
    1 * 60 件