Modules, Nunc-Immuno™ LockWell™
Product Details & Documents
PS. Nunc-Immuno™ LockWell™ modules facilitate assays from large scale screening to single-well tests. Available with a choice of MaxiSorp™, PolySorp™, MediSorp™ and MultiSorp™ surfaces. All MaxiSorp™ modules are certified for binding homogeneity. Framed modules (external dimensions: 128×86 mm) fit all standard equipment.
- Each well locks in the frame at the same height to prevent jamming in automated systems
- Strips are easy to take out or put in the frame, without use of any special tools
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains; PolySorp™ surface has a high affinity for hydrophobic molecules; MediSorp™ has a surface chemistry intermediate between PolySorp™ and MaxiSorp™, providing low background signal with samples containing serum; MultiSorp™ surfaces have a high affinity to hydrophilic molecules.
Colour coded modules available on request, please visit website or contact your local Avantor sales office for further information.
Documents
规格
- 孔数量:96
- 颜色:透明
- 说明:MaxiSorp™, U8×12
- 顺序:06
- Cat. No.:NUNC446639
- 环境认证:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact
Accessories
产品信息
- 目录编号顺序说明可用库存Price