Plates, Nunc™ OmniTrays
OmniTray with lid, +
目录编号 NUNC264728
Jump to:
Product Details & Documents
PS, sterile, with lid. Useful for screening libraries and for DNA hybridisation methods, these 90 ml trays are also suitable for standard Petri dish techniques and bacterial growth.
- Compatible with robotic systems
- Sturdy wall design facilitates handling and stacking
- Can be frozen at −20 °C
External dimensions: 128×86 mm
Recommended working volume: 35 ml
Recommended working volume: 35 ml
规格
- 灭菌:+
- 说明:OmniTray with lid
- Cat. No.:NUNC264728
- 环境认证:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact