比重计,M100系列
05, 1,200 - 1,300 g/ml, 0,002 g/ml, 250 mm
VWRI614-0113
不带温度计的玻璃比重计,用一个结实的透明塑料箱进行包装。
- Conform to ISO 649 (BS 718, DIN 12791, NF B 35511)
- Calibration temperature: 20 °C
- Division 0.002 g/ml
Length: 250 mm
: ISO 649, BS 718, DIN 12791, NF B 35511.
: In a strong transparent plastic case.
- 长度:250 mm
- 顺序:05
- 分度:0,002 g/ml
- 范围:1,200 - 1,300 g/ml