块状砝码,铁,M2级
04, 50 kg, 7500 ± mg
Catalog # VWRI611-8390
Supplier: VWR Collection
Jump to:
Product Details & Documents
矩形砝码,采用铸铁制成,永磁体。
- 不可拭除的独特识别编码确保可追溯性
- 灰色涂层
- 调整腔,以铅塞密封
符合OILM,COFRAC认证。
Certifications: Conforms to OIML, COFRAC accredited.
Documents
Specifications
- 重量:50 kg
- 顺序:04
- 误差:7500 ± mg