VWR®, Calibration Weights, Class M2
16, 10 kg, 1600 ± mg
目录编号 VWRI611-8382
供应商: VWR Collection
Jump to:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
规格
- 重量:10 kg
- 顺序:16
- 误差:1600 ± mg