VWR®, Calibration Weights, Class M2
20, 1 g, 3 ± mg
目录编号 VWRI611-8353
供应商: VWR Collection
Jump to:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
规格
- 重量:1 g
- 顺序:20
- 误差:3 ± mg