Separating funnels, spherical, DURAN®
2500 ml
目录编号 SCOT242916605
供应商: DWK LIFE SCIENCES
Jump to:
Product Details & Documents
DURAN® borosilicate glass 3.3, clear, stopcock with glass key and plastic stopper.
- Without graduation
- Very good chemical resistance
- Very good temperature resistance
规格
- 容量:2500 ml