订单输入
China
Orders LinkContactUsLinkComponent
 

Flasks, conical, Erlenmeyer shape, wide neck, DURAN®

5000 ml, 322 mm, 220 mm, 108 mm
目录编号 SCOT212277306
供应商:   DWK LIFE SCIENCES
Flasks, conical, Erlenmeyer shape, wide neck, DURAN®
目录编号 SCOT212277306
供应商:  DWK LIFE SCIENCES
Flasks, conical, Erlenmeyer shape, wide neck, DURAN®
Flasks, conical, Erlenmeyer shape, wide neck, DURAN®

5000 ml, 322 mm, 220 mm, 108 mm

Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP

With beaded rim and labelling area. Without graduations.

规格
  • 外径:
    220 mm
  • 容量:
    5000 ml
  • 高度:
    322 mm
  • 管颈外径:
    108 mm