Orders LinkContactUsLinkComponent
Multiple adapters
04, 29/32 NS, 29/32 NS
  LENZ05000444
Multiple adapters
  LENZ05000444
Multiple adapters
Multiple adapters
04, 29/32 NS, 29/32 NS
1 * 1 件
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two standard ground 29/32 sockets and one standard ground 29/32 cone. Side neck inclined upwards at an angle of 105°.
  • 圆锥体大小:
    29/32 NS
  • 插座尺寸:
    29/32 NS
  • 顺序:
    04