Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
500 ml, 29/32 NS, -
Catalog # LENZ04000058
Supplier:   Lenz Laborglas
Conical separating funnels
Catalog # LENZ04000058
Supplier:  Lenz Laborglas
Conical separating funnels
Conical separating funnels
500 ml, 29/32 NS, -
Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.
Documents
Specifications
  • 插座尺寸:
    29/32 NS
  • 容量:
    500 ml
  • 分度:
    -