硝酸锌六水合物 98%, extra pure
4024, Plastic bottle, 1 kg
硝酸锌六水合物 98%, extra pure
目录编号 ACRO211660010
CAS号: 10196-18-6
Jump to:
Product Details & Documents
同义词: Zinc nitrate hexahydrate
- CAS号:10196-18-6
- 化学式:ZnN₂O₆·6H₂O
- MDL 编号:MFCD00149889
- Molecular Weight:297.49 g/mol
- 默克索引:12,10273
- 沸点:? 105 °C (1013 hPa)
- 熔点:36 °C
- 密度:2,06 g/cm³ (20 °C)
- 储存温度:Ambient
- EINECS:231-943-8
- UN:1514
- ADR:5.1,II
Documents
规格
规格测试结果
- Appearance:White Adhering crystals
- Titration Complexometric:≥97.5 %
- Chloride (Cl):≤100 ppm
- Iron (Fe):≤20 ppm
- Lead (Pb):≤100 ppm
规格
- 包装类型:Plastic bottle
- 包装规格:1 kg
- 顺序:4024



