订单输入
China
Orders LinkContactUsLinkComponent
羟胺盐酸盐 97%

羟胺盐酸盐 97%

3401, Plastic bottle, 100 g
Catalog # ACRO170361000
Corrosive
Toxic
Health
Environment
羟胺盐酸盐 97%
Catalog # ACRO170361000
CAS Number:  5470-11-1
羟胺盐酸盐 97%
羟胺盐酸盐 97%

3401, Plastic bottle, 100 g

CAS Number:  5470-11-1
Price
Inquire For Price
Log in for availability
1 * 100 克
Product Details & Documents
Synonyms:  盐酸羟胺
  • CAS Number:5470-11-1
  • Formula:NH₂OH·HCl
  • MDL Number:MFCD00051089
  • Molecular Weight:69.49 g/mol
  • Merck Index:13,04853
  • Boiling Pt:306 °C (1013 hPa)
  • Melting Pt:155…158 °C (dec.)
  • Density:1,67 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:226-798-2
  • UN:2923
  • ADR:8,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    151 to 157 °C (Dec)
  • Titration with NaOH:
    ≥96.0 %

Specifications

  • 包装类型:
    Plastic bottle
  • 包装规格:
    100 g
  • 顺序:
    3401
  • Cat. No.:
    ACRO170361000

Specifications