乙醛肟,顺式和反式混合物 99%
3401, Glass bottle, 100 g
Jump to:
Product Details & Documents
同义词: 亚乙基羟胺
- CAS号:107-29-9
- 化学式:C₂H₅NO
- MDL 编号:MFCD00002124
- Molecular Weight:59.07 g/mol
- 熔点:44…46 °C
- 密度:0,96 g/cm³
- 闪点:38 °C
- 储存温度:Refrigerator
- EINECS:203-479-6
- UN:2332
- ADR:3,III
Documents
ACRO10223
SDS
规格
规格测试结果
- Appearance:Clear colorless to white Crystals or low melting solid or liquid
- Infrared spectrum:Conforms
- Refractive index:1.4230 to 1.4270 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤10 APHA
规格
- 包装类型:Glass bottle
- 包装规格:100 g
- 顺序:3401



