乙醛肟,顺式和反式混合物 99%
3308, Glass bottle, 25 g
: 亚乙基羟胺
- :107-29-9
- :C₂H₅NO
- :MFCD00002124
- :59.07 g/mol
- :44…46 °C
- :0,96 g/cm³
- :38 °C
- :Refrigerator
- :203-479-6
- :2332
- :3,III
Documents
ACRO10223
SDS
- Appearance:Clear colorless to white Crystals or low melting solid or liquid
- Infrared spectrum:Conforms
- Refractive index:1.4230 to 1.4270 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤10 APHA
- 包装类型:Glass bottle
- 包装规格:25 g
- 顺序:3308



