Affinity chromatography columns, HiTrap™ rProtein A FF
Product Details & Documents
HiTrap rProtein A FF columns are prepacked with rProtein A Sepharose Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources, including cell supernatants and ascites.
- Quick, convenient preparative purification of monoclonal antibodies
- Well-established protein A-based resin used in many approved MAb processes
- Simple operations with a syringe, pump, or high-performance chromatography system, such as ÄKTA design
Recombinant protein A exhibits similar Fc region specificity to that of native protein A but shows enhanced binding capacity and fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification.
The recombinant protein A ligand (produced in E. coli) is coupled to Sepharose Fast Flow by a technique which generates a stable thioether linkage between rProtein A and the base matrix. The coupling technique is optimized to give high binding capacity for IgG.
规格
- 流速:20 ml/min
- 颗粒大小:90 µm
- 容量:1×5 ml
- 说明:HiTrap™ rProtein A FF
- 顺序:04
- 硬件栏:PP
- Bed dimensions:16×25 mm
- Cat. No.:GEHE17-5080-01
- pH 范围:2 - 9
- 存储:4 to 30 °C, 20% Ethanol
- 最大压力:5 bar
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



