酞菁锌 96%
3173, Glass bottle, 5 g
Jump to:
Product Details & Documents
同义词: 酞菁
- CAS号:14320-04-8
- 化学式:C₃₂H₁₆N₈Zn
- MDL 编号:MFCD00041955
- Molecular Weight:577.91 g/mol
- 储存温度:Ambient
- EINECS:238-262-5
Documents
ACRO42275
SDS
规格
规格测试结果
- Appearance:Blue to purple Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 % (105°C)
- UV spectrophotometry:≥95 %
规格
- 包装类型:Glass bottle
- 包装规格:5 g
- 顺序:3173



