Buckets
Bucket, with graduation and handle, 白色, 10 L, 300 mm
BRND71774
: BRAND
HDPE, white.
- Good chemical and temperature resistance
- Scale with 1 litre steps
- Without spout and lid (Please order push-on lid seperately)
For industrial and laboratory applications. With handle.
- 容量:10 L
- 颜色:白色
- 说明:Bucket, with graduation and handle
- 高度:300 mm