Volumetric flasks, class B
1000 ml, 24/29, 0,8 ± ml
Catalog # BRND36062
Supplier: BRAND
Jump to:
Product Details & Documents
High clarity PP, with PP stopper.
- Calibrated to contain (TC, In)
- Ring mark and inscriptions in blue
- Exposure to temperatures up to +60 °C (autoclaving) will not cause permanently exceeded error limits, Cleaning up to max. +60 °C is recommended to preserve marks and inscriptions
Error limits according to class B, DIN EN ISO 1042.
*Wide neck.
Documents
Brand Volumetric Measurement in the Laboratory
Product Literature
BRND36020_TDS_EN BRND36028_TDS_EN
Datasheet
Specifications
- 模拟器:24/29
- 容量:1000 ml
- 误差:0,8 ± ml