DL-二硫苏糖醇
3173, Glass bottle for solids, 5 g
目录编号 BRAD161-0611
Marketsource
供应商: BIO RAD
Jump to:
Product Details & Documents
同义词: 1,4-二巯基苏糖醇, 克莱兰氏试剂, DTT
- CAS号:3483-12-3
- 化学式:C₄H₁₀O₂S₂
- MDL 编号:MFCD00004877
- Molecular Weight:154.25 g/mol
- 沸点:∼125 °C (1013 hPa)
- 熔点:42…44 °C
- 闪点:>110 °C
- 储存温度:Refrigerator
- EINECS:222-468-7
规格
规格
- 包装类型:Glass bottle for solids
- 包装规格:5 g
- 顺序:3173



