Scoops
1500 ml, 350 mm
目录编号 BOCH8803
供应商: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Designed for rough handling, anti-magnetic
规格
- 容量:1500 ml
- 长度:350 mm