Scoops
1000 ml, 320 mm
目录编号 BOCH8802
供应商: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Designed for rough handling, anti-magnetic
规格
- 容量:1000 ml
- 长度:320 mm