Scoops
450 ml, 250 mm
Catalog # BOCH8801
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Designed for rough handling, anti-magnetic
Specifications
- 容量:450 ml
- 长度:250 mm