Scoops
120 ml, 190 mm
目录编号 BOCH8800
供应商: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Designed for rough handling, anti-magnetic
规格
- 容量:120 ml
- 长度:190 mm